ENA048304198 Enamine

3-(1-ethyl-5-sulfamoyl-1H-1,3-benzodiazol-2-yl)propanoic acid

  • MDL number MFCD06655247
  • Cas Number : 847782-00-7
  • Molecular Weight : 297.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11737
Availability details for
Quantity
  • Availability
  • Add to Cart