ENA048304199 Enamine

1-(3,4-dipropoxyphenyl)ethan-1-amine

  • MDL number MFCD06373949
  • Cas Number : 105321-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11739
Availability details for
Quantity
  • Availability
  • Add to Cart