ENA048304209 Enamine

2-[3-chloro-2-oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]acetic acid

  • MDL number MFCD06375941
  • Cas Number : 851208-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-187
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11759

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11759
Availability details for
Quantity
  • Availability
  • Add to Cart