ENA048304216 Enamine

3-amino-N-(tert-butyl)benzenesulfonamide

  • MDL number MFCD06660528
  • Cas Number : 608523-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 137...139
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11771

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11771
Availability details for
Quantity
  • Availability
  • Add to Cart