ENA048304217 Enamine

2-(chloromethyl)-5-(3-chlorophenyl)-1,3-oxazole

  • MDL number MFCD06373945
  • Cas Number : 64640-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11772
Availability details for
Quantity
  • Availability
  • Add to Cart