ENA048304219 Enamine

5-[(2-methylprop-2-enyl)thio]-1,3,4-thiadiazole-2-thiol

  • MDL number MFCD06375942
  • Cas Number : 851208-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11775

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11775
Availability details for
Quantity
  • Availability
  • Add to Cart