ENA048304221 Enamine

1-(2,3-dimethylcyclohexyl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06375935
  • Cas Number : 953807-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-118
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11780

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11780
Availability details for
Quantity
  • Availability
  • Add to Cart