ENA048304229 Enamine

1-(3-methoxyphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD06375925
  • Cas Number : 51581-49-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11794
Availability details for
Quantity
  • Availability
  • Add to Cart