ENA048304231 Enamine

3-chloro-N-methyl-5-(trifluoromethyl)pyridin-2-amine

  • MDL number MFCD06375956
  • Cas Number : 89810-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 44...46
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11806

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11806
Availability details for
Quantity
  • Availability
  • Add to Cart