ENA048304235 Enamine

3,4-dihydronaphthalene-2-sulfonyl chloride

  • MDL number MFCD06655248
  • Cas Number : 17070-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11816
Availability details for
Quantity
  • Availability
  • Add to Cart