ENA048304241 Enamine

4-chloro-2-[(2-methoxyphenyl)amino]-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD06378223
  • Cas Number : 851288-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11827

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11827
Availability details for
Quantity
  • Availability
  • Add to Cart