ENA048304242 Enamine

1-(3,5-dimethylphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD06378222
  • Cas Number : 475997-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11828

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11828
Availability details for
Quantity
  • Availability
  • Add to Cart