ENA048304243 Enamine

{[1-(3,4-dimethylphenyl)-1H-imidazol-2-yl]thio}acetic acid

  • MDL number MFCD06378224
  • Cas Number : 851288-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11830

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11830
Availability details for
Quantity
  • Availability
  • Add to Cart