ENA048304247 Enamine

3-cyclopropyl-1-phenyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06378233
  • Cas Number : 890593-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11837

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11837
Availability details for
Quantity
  • Availability
  • Add to Cart