ENA048304248 Enamine

N-[2-(dimethylamino)ethyl]-2-hydroxybenzamide

  • MDL number MFCD06379298
  • Cas Number : 91430-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11842
Availability details for
Quantity
  • Availability
  • Add to Cart