ENA048304249 Enamine

6-cyclopropyl-1-(propan-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD06378235
  • Cas Number : 851288-57-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11844
Availability details for
Quantity
  • Availability
  • Add to Cart