ENA048304257 Enamine

1-[(4-bromothiophen-2-yl)methyl]-1H-pyrazol-5-amine

  • MDL number MFCD06378269
  • Cas Number : 956453-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11866
Availability details for
Quantity
  • Availability
  • Add to Cart