ENA048304264 Enamine

2-chloro-N-(3-methoxybenzyl)-N-methylacetamide

  • MDL number MFCD06373947
  • Cas Number : 851176-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11900

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11900
Availability details for
Quantity
  • Availability
  • Add to Cart