ENA048304279 Enamine

1-(1-chloroethyl)-4-fluorobenzene

  • MDL number MFCD06376752
  • Cas Number : 456-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11932

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-11932
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart