ENA048304282 Enamine

5-ethyl-2-mercapto-3-(4-methoxy-2-methylphenyl)-6-methylthieno[2,3-d]pyrimidin-4(3H)-one

  • MDL number MFCD06655254
  • Cas Number : 854039-11-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11948

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11948
Availability details for
Quantity
  • Availability
  • Add to Cart