ENA048304294 Enamine

5-{[(1-carboxy-2-methylbutyl)amino]sulfonyl}-2-chlorobenzoic acid

  • MDL number MFCD06257151
  • Cas Number : 1008040-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11733

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11733
Availability details for
Quantity
  • Availability
  • Add to Cart