ENA048304300 Enamine

1-[2-(4-methoxyphenoxy)ethyl]piperazine

  • MDL number MFCD03231419
  • Cas Number : 117132-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31...33
Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11762

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11762
Availability details for
Quantity
  • Availability
  • Add to Cart