ENA048304301 Enamine

4-oxo-1H,2H,3H,3aH,4H,5H-pyrrolo[1,2-a]quinoxaline-7-carboxylic acid

  • MDL number MFCD04124142
  • Cas Number : 1008665-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 250-251
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11838
Availability details for
Quantity
  • Availability
  • Add to Cart