ENA048304318 Enamine

2-{[3-cyano-4-(furan-2-yl)-5,6,7,8-tetrahydroquinolin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD00408509
  • Cas Number : 94640-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11788
Availability details for
Quantity
  • Availability
  • Add to Cart