ENA048304331 Enamine

2-amino-5-(propan-2-yl)-4,5-dihydro-1,3-oxazol-4-one

  • MDL number MFCD06378273
  • Cas Number : 15900-26-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11868
Availability details for
Quantity
  • Availability
  • Add to Cart