ENA048304360 Enamine

2-(N-benzylbenzenesulfonamido)acetic acid

  • MDL number MFCD00532367
  • Cas Number : 276695-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11897
Availability details for
Quantity
  • Availability
  • Add to Cart