ENA048304361 Enamine

6-ethyl-7-hydroxy-4-methyl-2H-chromen-2-one

  • MDL number MFCD01106609
  • Cas Number : 1484-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11898
Availability details for
Quantity
  • Availability
  • Add to Cart