ENA048304363 Enamine

N-(1-phenylethyl)-N-(pyridin-4-ylmethyl)amine

  • MDL number MFCD01475086
  • Cas Number : 289476-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11801

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11801
Availability details for
Quantity
  • Availability
  • Add to Cart