ENA048304373 Enamine

4-(1,3-benzothiazol-2-yl)phenol

  • MDL number MFCD01707773
  • Cas Number : 6265-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11910
Availability details for
Quantity
  • Availability
  • Add to Cart