ENA048304374 Enamine

2-chloro-N-methyl-N-({[2-(trifluoromethyl)phenyl]carbamoyl}methyl)acetamide

  • MDL number MFCD05266748
  • Cas Number : 744227-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11911
Availability details for
Quantity
  • Availability
  • Add to Cart