ENA048304375 Enamine

2-[N-(2-methylphenyl)methanesulfonamido]acetic acid

  • MDL number MFCD02219537
  • Cas Number : 362695-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-110
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11912
Availability details for
Quantity
  • Availability
  • Add to Cart