ENA048304381 Enamine

5-methyl-4,5,6,7-tetrahydro-1-benzothiophene-2-carbohydrazide

  • MDL number MFCD04969093
  • Cas Number : 777880-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122...124
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11918

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11918
Availability details for
Quantity
  • Availability
  • Add to Cart