ENA048304396 Enamine

4-(4-ethoxyphenyl)-5-(4-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD06368752
  • Cas Number : 380307-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11933
Availability details for
Quantity
  • Availability
  • Add to Cart