ENA048304406 Enamine

2,6-dimethyl-4-(morpholin-4-ylmethyl)phenol

  • MDL number MFCD06357131
  • Cas Number : 21894-01-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 185...187
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11943
Availability details for
Quantity
  • Availability
  • Add to Cart