ENA048304407 Enamine

N-benzyl-4-methylaniline

  • MDL number MFCD00870620
  • Cas Number : 5405-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11944
Availability details for
Quantity
  • Availability
  • Add to Cart