ENA048304421 Enamine

4-(9H-fluoren-3-yl)-1,3-thiazol-2-amine

  • MDL number MFCD05511009
  • Cas Number : 885460-95-7
  • Molecular Weight : 264.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(9H-fluoren-3-yl)-1,3-thiazol-2-amine
Inchi Code 1S/C16H12N2S/c17-16-18-15(9-19-16)12-6-5-11-7-10-3-1-2-4-13(10)14(11)8-12/h1-6,8-9H,7H2,(H2,17,18)
Inchi Key LTQJZSRNRYWUSV-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11958
Availability details for
Quantity
  • Availability
  • Add to Cart