ENA053246015 Enamine

1-butyl-3-phenyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD06655013
  • Cas Number : 901273-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-08012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08012
Availability details for
Quantity
  • Availability
  • Add to Cart