ENA053246025 Enamine

2-(trifluoromethyl)-1,3-oxathiolan-5-one

  • MDL number MFCD06660750
  • Cas Number : 79413-31-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11970
Availability details for
Quantity
  • Availability
  • Add to Cart