ENA053246029 Enamine

4-Chloro-3-trifluoromethyl-benzenethiol

  • MDL number MFCD00068898
  • Cas Number : 18800-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148...150
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12128

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12128
Availability details for
Quantity
  • Availability
  • Add to Cart