ENA053246031 Enamine

2,5-dichloro-3,6-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD06655261
  • Cas Number : 854860-52-9
  • Molecular Weight : 273.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-56
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-12161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12161
Availability details for
Quantity
  • Availability
  • Add to Cart