ENA053246033 Enamine

ethyl (tert-butylthio)acetate

  • MDL number MFCD06660517
  • Cas Number : 60178-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12171

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12171
Availability details for
Quantity
  • Availability
  • Add to Cart