ENA053246036 Enamine

2,3-dihydro-1H-inden-5-yl hydrosulfide

  • MDL number MFCD06655306
  • Cas Number : 55119-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12348
Availability details for
Quantity
  • Availability
  • Add to Cart