ENA053246047 Enamine

5-bromo-2-morpholin-4-ylaniline

  • MDL number MFCD06660760
  • Cas Number : 91062-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153...155
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12395

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12395
Availability details for
Quantity
  • Availability
  • Add to Cart