ENA053246057 Enamine

5-acetamido-2-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD06660768
  • Cas Number : 13632-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-125
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12409

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12409
Availability details for
Quantity
  • Availability
  • Add to Cart