ENA053246072 Enamine

N-cyclopropyl-1,3-benzothiazol-2-amine

  • MDL number MFCD06660784
  • Cas Number : 885461-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12444

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12444
Availability details for
Quantity
  • Availability
  • Add to Cart