ENA053246083 Enamine

3,6-dimethyl-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylic acid

  • MDL number MFCD06660795
  • Cas Number : 885461-44-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12464

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12464
Availability details for
Quantity
  • Availability
  • Add to Cart