ENA053246107 Enamine

2-[2,4,5-trioxo-3-(prop-2-en-1-yl)imidazolidin-1-yl]acetic acid

  • MDL number MFCD06660819
  • Cas Number : 885461-65-4
  • Molecular Weight : 212.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12500

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12500
Availability details for
Quantity
  • Availability
  • Add to Cart