ENA053246108 Enamine

2-(4-chlorophenyl)-5-methyl-1,3-oxazole-4-carboxylic acid

  • MDL number MFCD06660820
  • Cas Number : 2940-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12501

Safety Information

msds https://www.enaminestore.com/catalog/EN300-12501
Availability details for
Quantity
  • Availability
  • Add to Cart