ENA053246120 Enamine

4-chloro-1-(3,5-dimethylphenyl)-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD06660833
  • Cas Number : 885524-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12896
Availability details for
Quantity
  • Availability
  • Add to Cart