ENA053246124 Enamine

1-[2-(propan-2-yl)phenyl]-1H-imidazole-2-thiol

  • MDL number MFCD06382007
  • Cas Number : 25372-32-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-12052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-12052
Availability details for
Quantity
  • Availability
  • Add to Cart